missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPL39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RPL39 |
---|---|
Concentration | 0.2 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16883944
|
Proteintech
14990-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16873944
|
Proteintech
14990-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ribosomes, the organelles that catalyze protein synthesis, are composed of a small subunit (40S) and a large subunit (60S) that consist of over 80 distinct ribosomal proteins. Mammalian ribosomal proteins are encoded by multigene families that contain processed pseudogenes and one functional intron-containing gene within their coding regions. Ribosomal Protein L39 (RPL39), also known as 60S Ribosomal Protein L39, consists of 51 amino acids and is the smallest protein found in liver ribosomes. Ribosomal Protein L39 localizes to cytoplasm and is a member of the S39E family of ribosomal proteins. The gene encoding Ribosomal Protein L39 maps to human chromosome Xq24, and shares 92 percent homology with Ribosomal Protein L39L (Ribosomal Protein L39-like), which is also known as RPL39L, L39-2 or RPL39L1. Ribosomal Protein L39L is associated with the 60S ribosomal subunit and is expressed in testis with localization in nucleolus.Specifications
RPL39 | |
Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P62891, P62892, P62893 | |
25347, 6170, 67248 | |
RPL39 Fusion Protein Ag6975 | |
Primary | |
-20°C | |
RPL39 |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
60S ribosomal protein L39, ribosomal protein L39, RPL39 | |
RPL39 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPL39 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech