Learn More
RPS14 Mouse anti-Human, Unconjugated, Clone: 3D5H5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67566-1-IG-20UL
 View more versions of this product
Product Code. 16880266
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The RPS14 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RPS14. This antibody recognizes human, mouse, rat antigen. The RPS14 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
RPS14
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
3D5H5
Unconjugated
20 μL
6208
IgG2b
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPS14 Mouse anti-Human, Unconjugated, Clone: 3D5H5, Proteintech > 20 μL