missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RPS19 Recombinant Rabbit Monoclonal Antibody (JU33-43), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA534641
This item is not returnable.
View return policy
Description
Positive Control: CRC, MCF-7, rat testis tissue, human colon cancer tissue, mouse colon tissue, mouse brain tissue, rat brain tissue.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S19E family of ribosomal proteins. It is located in the cytoplasm. Mutations in this gene cause Diamond-Blackfan anemia, a constitutional erythroblastopenia characterized by absent or decreased erythroid precursors, in a subset of patients. This suggests a possible extra-ribosomal function for this gene in erythropoietic differentiation and proliferation, in addition to its ribosomal function. Higher expression levels of this gene in some primary colon carcinomas compared to matched normal colon tissues has been observed. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
RPS19 | |
Monoclonal | |
1 mg/mL | |
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P39019 | |
RPS19 | |
Synthetic peptide within Human RPS19 aa 96-145 | |
100 ÎĽL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
JU33-43 | |
Unconjugated | |
RPS19 | |
40S ribosomal protein S19; DBA; DBA1; Dsk3; HGNC:10402; ribosomal protein S19; Rps19; S19; Small ribosomal subunit protein eS19 | |
Rabbit | |
Protein A | |
RUO | |
6223 | |
-20° C, Avoid Freeze/Thaw Cycles, store in dark | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RPS19 Recombinant Rabbit Monoclonal Antibody (JU33-43), Invitrogen™ > 100 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction