Learn More
RSAD2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28089-1-AP-20UL
 View more versions of this product
Product Code. 16885985
  • 1645.54 SEK / 20µL
Description
The RSAD2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RSAD2. This antibody recognizes human, mouse antigen. The RSAD2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
RSAD2
Polyclonal
Unconjugated
20 μL
91543
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
400 μg/mL
Rabbit
Primary
Human, Mouse
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
RSAD2 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech > 20 μL