missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RSPO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17193-1-AP-20UL
This item is not returnable.
View return policy
Description
RSPO3 is a member of the R-spondin family and plays a role in the regulation of Wnt (wingless-type MMTV integration site family)/beta-catenin and Wnt/planar cell polarity (PCP) signaling pathways, which are involved in development, cell growth and disease pathogenesis. RSPO3 has been shown to be involved in regulating the balance between angioblast and blood cell specification during embryonic vasculogenesis and angiogenesis and may be involved in tumor development.Specifications
RSPO3 | |
Polyclonal | |
Unconjugated | |
RSPO3 | |
CRISTIN1, hPWTSR, hRspo3, Protein with TSP type 1 repeat, PWTSR, R spondin 3, Roof plate specific spondin 3, RSPO3, THSD2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
498997, 72780, 84870 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q2TJ95, Q9BXY4 | |
Rspo3 | |
RSPO3 Fusion Protein Ag9899 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RSPO3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction