missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNDC3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | RUNDC3A |
---|---|
Concentration | 0.35 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890075
|
Proteintech
20531-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880075
|
Proteintech
20531-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May act as an effector of RAP2A in neuronal cells.Specifications
RUNDC3A | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Rat, Mouse | |
O08576, Q59EK9 | |
10900, 303569, 51799 | |
RUNDC3A Fusion Protein Ag14393 | |
Primary | |
-20°C | |
RUNDC3A |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Rap2 interacting protein 8, RAP2IP, RPIP 8, RPIP8, RUN domain containing 3A, RUNDC3A | |
Rundc3a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUNDC3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech