missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RUNX1T1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15494-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the Ser/Thr protein kinase family. This kinase functions as a mitogen-activated protein kinase (MAP kinase)- activated protein kinase. MAP kinases are also known as extracellular signal-regulated kinases (ERKs), act as an integration point for multiple biochemical signals. This kinase was shown to be activated by growth inducers and stress stimulation of cells. In vitro studies demonstrated that ERK, p38 MAP kinase and Jun N-terminal kinase were all able to phosphorylate and activate this kinase, which suggested the role of this kinase as an integrative element of signaling in both mitogen and stress responses. This kinase was reported to interact with, phosphorylate and repress the activity of E47, which is a basic helix-loop-helix transcription factor known to be involved in the regulation of tissue-specific gene expression and cell differentiation.Specifications
RUNX1T1 | |
Polyclonal | |
Unconjugated | |
RUNX1T1 | |
AML1T1, CBFA2T1, CDR, Cyclin D related protein, Eight twenty one protein, ETO, MTG8, MTG8b, Protein CBFA2T1, Protein ETO, Protein MTG8, RUNX1T1, ZMYND2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12395, 862 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q06455, Q61909 | |
RUNX1T1 | |
RUNX1T1 Fusion Protein Ag7892 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUNX1T1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction