Learn More
Emma Safety Footwear Ryan S3 Shoe
The Ryan has everything you would expect from a safety boot; rugged and durable with all day long comfort and protection for your feet in all conditions.
Brand:  Emma Safety Footwear MM73454846
Additional Details : Weight : 1.27000kg
 View more versions of this product
Product Code. 17522526
  • 1224.30 SEK / 1 pair
Estimated Shipment: 12-06-2024
to see stock.
Add to basket
Description
Description
  • Waterproof
  • The all-round safety boot: extremely safe & comfortable, 100% metal free & waterproof.
  • The Ryan has everything you would expect from a safety boot; rugged and durable with all day long comfort and protection for your feet in all conditions.
  • This boot has a fully waterproof membrane, so you don't have to worry about wet feet. You can fully concentrate on getting the job done.
  • The reflective parts on the boot ensure that you are clearly visible in the dark and the Gripforce outsole gives you maximum grip.
  • The extremely cool carbon look on the side of the upper gives the boot a sporty look.
  • It is available in 2 widths (D & XD).
  • Industries
    • Chemical industry
    • Metal industry
    • Construction & technology
    • Manufacturing
    • Nature & environment
  • Safety shoe
    • S3-Safetyshoe with sole plate for wet work
Specifications
Specifications
Laces
Hydro-Tec Antibacterial/DryShield membrane
46
Standard Width
S3
Black
Leather
ISO 20345:2011