missing translation for 'onlineSavingsMsg'
Learn More
(S)-1-Boc-2-isopropylpiperazine, 97% Product Code.: 15449157

(S)-1-Boc-2-isopropylpiperazine, 97%

Product Code. p-7034726
250 mg, 250mg
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15449157

Brand: Thermo Scientific Alfa Aesar H52424.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 674792-05-3 | C12H24N2O2 | 228.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 674792-05-3
Molecular Formula C12H24N2O2
Molecular Weight (g/mol) 228.34
MDL Number MFCD03787918
InChI Key NZTWGWFHWJARJX-SNVBAGLBSA-N
Synonym s-1-boc-2-isopropylpiperazine, s-1-boc-2-isopropyl-piperazine, s-tert-butyl 2-isopropylpiperazine-1-carboxylate, tert-butyl 2s-2-propan-2-yl piperazine-1-carboxylate, 1-n-boc-2s-isopropyl-piperazine, tert-butyl 2s-2-isopropylpiperazine-1-carboxylate, s-n-1-boc-2-isopropylpiperazine, 2s-1-n-boc-2-isopropylpiperazine, s-1-n-boc-2-isopropyl piperazineShow MoreShow Less
PubChem CID 17750439
IUPAC Name tert-butyl (2S)-2-propan-2-ylpiperazine-1-carboxylate
SMILES CC(C)[C@H]1CNCCN1C(=O)OC(C)(C)C

Specifications

Quantity 250 mg
Sensitivity Moisture sensitive
Formula Weight 228.33
Percent Purity 97%
Chemical Name or Material (S)-1-Boc-2-isopropylpiperazine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot NumberJ26W002 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH52424.03,H52424.MD
Lot NumberZ19D037 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH52424.03,H52424.MD
Lot NumberN29C017 Certificate TypeCertificate of Analysis Date07/12/2021 Product CodeH52424.03,H52424.MD

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-1-Boc-2-isopropylpiperazine, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.