Learn More
S100A8 Mouse anti-Human, Unconjugated, Clone: 1G11C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66853-1-IG-150UL
 View more versions of this product
Product Code. 16879105
  • 4652.90 SEK / 150µL
Explore more special offers
Description
Description
The S100A8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A8. This antibody recognizes human antigen. The S100A8 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
S100A8
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Immunohistochemistry, Immunofluorescence, ELISA
1G11C8
Unconjugated
150 μL
6279
IgG2a
Product Title
S100A8 Mouse anti-Human, Unconjugated, Clone: 1G11C8, Proteintech > 150 μL