Learn More
S100A8 Mouse anti-Human, Unconjugated, Clone: 1G11C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66853-1-IG-150UL
Description
The S100A8 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human S100A8. This antibody recognizes human antigen. The S100A8 antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
S100A8 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human |
Immunohistochemistry, Immunofluorescence, ELISA | |
1G11C8 | |
Unconjugated | |
150 μL | |
6279 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.