missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SALL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12679-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein containing multiple zinc finger domains. The encoded protein functions in optical fissure closure during development of the eye in the embryo. Mutations in this gene are associated with ocular coloboma.Specifications
SALL2 | |
Polyclonal | |
Unconjugated | |
SALL2 | |
HSAL2, KIAA0360, p150(Sal2), Sal 2, sal like 2 (Drosophila), Sal like protein 2, SAL2, SALL2, Zinc finger protein SALL2, Zinc finger protein Spalt 2, ZNF795 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
305854, 50524, 6297 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QX96, Q9Y467 | |
SALL2 | |
SALL2 Fusion Protein Ag3367 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SALL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction