missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-COV-2 3CL Protease (aa1-306), His and Avi Tag Recombinant Protein
E. coli Recombinant Protein
Brand: Invitrogen™ RP87698
Additional Details : Weight : 0.01000kg
Description
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has been attributed to a novel coronavirus (SARS-CoV). The SARS 3C-like protease (SARS 3CL (pro)) is a cysteine protease engaging in the proteolytic cleavage of the viral precursor polyprotein to a series of functional proteins required for corona virus replication and is considered an attractive target for therapeutics against SARS.- Purity: > 95% by SDS-PAGE
- Endotoxin Conentration: < 1.0 EU/μg of the protein by LAL method.
- Protein Length/ AA Sequence: Ser1-Gln306 of 2019-ncov 3C-like Proteinase fused with an initial Met, a 6×His, Avi tag at the N-terminus.
- Molecular Weight: 36.9 kDa
- Animal Origin Free: Yes Storage: This product is stable at -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature.Avoid repeated freeze-thaw cycles.
Specifications
Control, ELISA, Western Blot | |
36.9 kDa | |
100 μL | |
-80°C, Avoid Freeze/Thaw Cycles | |
Coronavirus; COVID; COVID 19; COVID-19; SARS Coronavirus; SARS CoV; SARS CoV2; SARS-CoV-2 | |
Unconjugated | |
Liquid |
20 mM HEPES with 10% glycerol, 120 mM NaCl, 2 mM TCEP and no preservative; pH 7.4 | |
SARS-COV-2 3 CL Protease (aa1-306), His and Avi Tag | |
E. coli | |
RUO | |
Virus | |
Recombinant | |
>95% |