missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SARS-COV-2 3CL Protease (aa1-306), His and Avi Tag Recombinant Protein
E. coli Recombinant Protein
4030.00 SEK - 33350.00 SEK
Specifications
For Use With (Application) | Control, ELISA, Western Blot |
---|---|
Formulation | 20 mM HEPES with 10% glycerol, 120 mM NaCl, 2 mM TCEP and no preservative; pH 7.4 |
Molecular Weight (g/mol) | 36.9 kDa |
Name | SARS-COV-2 3 CL Protease (aa1-306), His and Avi Tag |
Source | E. coli |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17173773
|
Invitrogen™
RP87698 |
100 μL |
4030.00 SEK
100µL |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17183773
|
Invitrogen™
RP87699 |
1 mg |
33350.00 SEK
1mg |
Estimated Shipment: 03-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has been attributed to a novel coronavirus (SARS-CoV). The SARS 3C-like protease (SARS 3CL (pro)) is a cysteine protease engaging in the proteolytic cleavage of the viral precursor polyprotein to a series of functional proteins required for corona virus replication and is considered an attractive target for therapeutics against SARS.- Purity: > 95% by SDS-PAGE
- Endotoxin Conentration: < 1.0 EU/μg of the protein by LAL method.
- Protein Length/ AA Sequence: Ser1-Gln306 of 2019-ncov 3C-like Proteinase fused with an initial Met, a 6×His, Avi tag at the N-terminus.
- Molecular Weight: 36.9 kDa
- Animal Origin Free: Yes Storage: This product is stable at -70°C for up to 1 year from the date of receipt. For optimal storage, aliquot into smaller quantities after centrifugation and store at recommended temperature.Avoid repeated freeze-thaw cycles.
Specifications
Control, ELISA, Western Blot | |
36.9 kDa | |
E. coli | |
RUO | |
Virus | |
Recombinant | |
>95% |
20 mM HEPES with 10% glycerol, 120 mM NaCl, 2 mM TCEP and no preservative; pH 7.4 | |
SARS-COV-2 3 CL Protease (aa1-306), His and Avi Tag | |
-80°C, Avoid Freeze/Thaw Cycles | |
Coronavirus; COVID; COVID 19; COVID-19; SARS Coronavirus; SARS CoV; SARS CoV2; SARS-CoV-2 | |
Unconjugated | |
Liquid |