Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (K417T, E484K, N501Y), His Tag (MALS verified)
Brand:  ACROBiosystems SPD-C52Hr-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17216530
  • 33904.71 SEK / 1mg
Estimated Shipment: 12-10-2023
to see stock.
Description
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 26.5 kDa. The protein migrates as 33-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
QHD43416.1 (K417T, E484K, N501Y)
1 mg
Arg 319 - Lys 537
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19
Unconjugated
His Tag
Less than 1.0 EU / µg by the LAL method.
Powder
>95% as determined by SDS-PAGE.
26.5 kDa
SARS-CoV-2 S protein RBD (K417T, E484K, N501Y), His Tag (SPD-C52Hr) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T, E484K, N501Y)). K417T/ E484K/ N501Y mutations were identified in the SARS-CoV-2 variant (known as variant P0.1) which emerged in Brazil.
-20°C
S protein RBD
SARS-CoV-2
PBS, pH7.4
HEK293
Viral Proteins