Learn More
ACROBiosystems SARS-CoV-2 S protein RBD (K417T, E484K, N501Y), His Tag (MALS verified)

Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
Specifications
Specifications
Accession Number | QHD43416.1 (K417T, E484K, N501Y) |
Molecular Weight (g/mol) | 26.5 kDa |
Quantity | 100 μg |
Source | SARS-CoV-2 S protein RBD (K417T, E484K, N501Y), His Tag (SPD-C52Hr) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (K417T, E484K, N501Y)). K417T/ E484K/ N501Y mutations were identified in the SARS-CoV-2 variant (known as variant P0.1) which emerged in Brazil. |
Immunogen | Arg 319 - Lys 537 |
Storage Requirements | -20°C |
Gene Alias | Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 |
Common Name | S protein RBD |
Conjugate | Unconjugated |
Species | SARS-CoV-2 |
Show More |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.