missing translation for 'onlineSavingsMsg'
Learn More
ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G), Fc Tag Product Code.: 17236440

ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G), Fc Tag

Product Code. 17236440
1 mg, 1mg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 ÎĽg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 17236440

Brand: ACROBiosystems S1DC52561mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101 kDa.

Specifications

Molecular Weight (g/mol) 101 kDa
Quantity 1 mg
Source SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1).
Immunogen Val 16 - Arg 685
Storage Requirements -20°C
Gene Alias Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Common Name S1 protein
Conjugate Unconjugated
Species SARS-CoV-2
Protein Tag Fc Tag
Expression System HEK293
Form Powder
Protein Subtype Viral Proteins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G), Fc Tag

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.