missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G), Fc Tag

5085.71 SEK - 36773.57 SEK

Specifications

Molecular Weight (g/mol) 101 kDa
Source SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1).
Immunogen Val 16 - Arg 685
Storage Requirements -20°C
Gene Alias Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17226440
View Documents Promotion Details
ACROBiosystems
S1D-C5256-100UG
100 μg
5085.71 SEK
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 12-10-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
17236440
View Documents Promotion Details
ACROBiosystems
S1D-C5256-1MG
1 mg
36773.57 SEK
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 12-10-2023
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101 kDa.
Specifications

Specifications

101 kDa
Val 16 - Arg 685
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19
Unconjugated
Fc Tag
Powder
SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1).
-20°C
S1 protein
SARS-CoV-2
HEK293
Viral Proteins
Videos
SDS
Documents

Documents

Product Certifications