Learn More
ACROBiosystems SARS-CoV-2 S1 protein (L18F, D80A, D215G, LAL242-244del, R246I, K417N, E484K, N501Y, D614G), Fc Tag
Brand: ACROBiosystems S1D-C5256-100ug
Additional Details : Weight : 0.00010kg
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 101 kDa.Specifications
Powder | |
100 μg | |
Val 16 - Arg 685 | |
Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1,COVID-19 | |
Unconjugated | |
HEK293 | |
SARS-CoV-2 |
101 kDa | |
SARS-CoV-2 S1 protein, Fc Tag (S1D-C5256) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1). | |
-20°C | |
S1 protein | |
Fc Tag | |
Viral Proteins |