Learn More
ACROBiosystems SARS-CoV-2 S2 protein (T1027I, V1176F), His Tag
Brand:  ACROBiosystems S2N-C52He-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 17216500
  • 29079.81 SEK / 1mg
Estimated Shipment: 13-10-2023
to see stock.
Description
Description
The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1,000,000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Mouse Anti-SARS-CoV-2 antibody IgG Serologic Assay kit.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.
Specifications
Specifications
59.8 kDa
SARS-CoV-2 S2 protein (T1027I, V1176F), His Tag (S2N-C52He) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (T1027I, V1176F, F817P, A892P, A899P, A942P, K986P, V987P)). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein.)
-20°C
S2 protein
SARS-CoV-2
HEK293
Viral Proteins
1 mg
Ser 686 - Pro 1213
Spike,S2 protein,Spike glycoprotein Subunit2,S glycoprotein Subunit2,Spike protein S2,COVID-19
Unconjugated
His Tag
Powder