missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems SARS-CoV-2 S2 protein (T1027I, V1176F), His Tag
4005.50 SEK - 29640.69 SEK
Specifications
Molecular Weight (g/mol) | 59.8 kDa |
---|---|
Source | SARS-CoV-2 S2 protein (T1027I, V1176F), His Tag (S2N-C52He) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (T1027I, V1176F, F817P, A892P, A899P, A942P, K986P, V987P)). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein.) |
Immunogen | Ser 686 - Pro 1213 |
Storage Requirements | -20°C |
Gene Alias | Spike,S2 protein,Spike glycoprotein Subunit2,S glycoprotein Subunit2,Spike protein S2,COVID-19 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17206500
|
ACROBiosystems
S2N-C52HE-100UG |
100 μg |
4005.50 SEK
100µg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17216500
|
ACROBiosystems
S2N-C52HE-1MG |
1 mg |
29640.69 SEK
1mg |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The ongoing SARS-COV-2 pandemics has sickened 3.7 million people around the world, claiming over 1,000,000 lives. To deal with this global public health crisis, unprecedented efforts are being made to study the virus, SARS-CoV-2. At present, there are 193 new crown vaccine projects registered with the World Health Organization are being developed. To support these studies, ACRO has developed Mouse Anti-SARS-CoV-2 antibody IgG Serologic Assay kit.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 59.8 kDa.Specifications
59.8 kDa | |
Ser 686 - Pro 1213 | |
Spike,S2 protein,Spike glycoprotein Subunit2,S glycoprotein Subunit2,Spike protein S2,COVID-19 | |
Unconjugated | |
His Tag | |
Powder |
SARS-CoV-2 S2 protein (T1027I, V1176F), His Tag (S2N-C52He) is expressed from human 293 cells (HEK293). It contains AA Ser 686 - Pro 1213 (Accession # QHD43416.1 (T1027I, V1176F, F817P, A892P, A899P, A942P, K986P, V987P)). The recombinant protein is expressed from human 293 cells (HEK293) with T4 fibritin trimerization motif and a polyhistidine tag (at the C-terminus. Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein.) | |
-20°C | |
S2 protein | |
SARS-CoV-2 | |
HEK293 | |
Viral Proteins |