Learn More
SARS-CoV-2 Spike Human anti-Virus, Unconjugated, Clone: AM006415, Proteintech
Human Recombinant Antibody
2258.36 SEK - 6809.13 SEK
Specifications
Antigen | SARS-CoV-2 Spike |
---|---|
Clone | AM006415 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890446
|
Proteintech
91351-PTG-20UG |
20 ÎĽg |
2258.36 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880446
|
Proteintech
91351-PTG-100UG |
100 ÎĽg |
6809.13 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The SARS-CoV-2 Spike antibody from Proteintech is a human recombinant antibody generated with virus of virus SARS-CoV-2 Spike. This antibody recognizes virus antigen. The SARS-CoV-2 Spike antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
SARS-CoV-2 Spike | |
Western Blot, ELISA | |
Unconjugated | |
Virus | |
IgG1 |
AM006415 | |
Monoclonal | |
Human | |
43740568 | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.