Learn More
Sartorius Sartoclear Dynamics™ Lab V Filtration Kit
Sartorius Sartoclear Dynamics™ Lab V Kits designed for the clarification and sterile filtration of mammalian cell culture with diatomaceous earth as filter aid.
Specifications
Product Type Vacuum Filtration Kit
View More Specs
Description
Description
Comprised of:
  • A graduated funnel with a polyethersulfone (PES) membrane, a vacuum adapter and a lid
  • A bottle, with graduation and writing field
  • A tube connector for vacuum connection (for stand-alone filtration)
  • A screw cap for storage of the filtrate (individually wrapped to maintain sterility) history
Benefits:
  • Kits for a single-step mammalian cell culture harvesting
  • Ready to use
  • Fast and effortless
Specifications
Specifications
Vacuum Filtration Kit
Documents
Product Certifications
Product Content Correction
Product Title
Sartorius Sartoclear Dynamics™ Lab V Filtration Kit