Learn More
SBDS Mouse anti-Human, Unconjugated, Clone: 1F8A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67200-1-IG-150UL
 View more versions of this product
Product Code. 16879705
  • 4652.90 SEK / 150µL
Description
The SBDS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SBDS. This antibody recognizes human, mouse, rat antigen. The SBDS antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SBDS
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1F8A10
Unconjugated
150 μL
51119
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SBDS Mouse anti-Human, Unconjugated, Clone: 1F8A10, Proteintech > 150 μL