Learn More
SBDS Mouse anti-Human, Unconjugated, Clone: 1F8A10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67200-1-IG-150UL
 View more versions of this product
Product Code. 16879705
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
The SBDS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SBDS. This antibody recognizes human, mouse, rat antigen. The SBDS antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
SBDS
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence
1F8A10
Unconjugated
150 μL
51119
IgG2a
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SBDS Mouse anti-Human, Unconjugated, Clone: 1F8A10, Proteintech > 150 μL