Läs mer
Scandium(III) bromide, ultra dry, 99.99% (metals basis), Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 047199.03
| Kvantitet | 1g |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ampouled under argon
Kemiska identifierare
| 13465-59-3 | |
| 284.67 | |
| APPHYFNIXVIIJR-UHFFFAOYSA-K | |
| 83495 | |
| [Sc+3].[Br-].[Br-].[Br-] |
| Br3Sc | |
| MFCD00016324 | |
| scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis | |
| scandium(3+) tribromide |
Specifikationer
| Scandium(III) bromide | |
| 13465-59-3 | |
| (metals basis) | |
| MFCD00016324 | |
| APPHYFNIXVIIJR-UHFFFAOYSA-K | |
| scandium(3+) tribromide | |
| 83495 | |
| 99.99% | |
| Hygroscopic |
| Ultra dry | |
| 1g | |
| Br3Sc | |
| scandium tribromide, scandium bromide scbr3, scandium iii bromide, scandium iii bromide hydrate, scandium iii bromide, anhydrous, powder trace metals basis | |
| [Sc+3].[Br-].[Br-].[Br-] | |
| 284.67 | |
| 284.67 | |
| Ampule |