Learn More
SCGBL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21142-1-AP-150UL
 View more versions of this product
Product Code. 16880745
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Specifications
SCGBL
Polyclonal
Unconjugated
SCGB2B2
SCGBL, secretoglobin like, Secretoglobin like protein
Rabbit
Antigen Affinity Chromatography
RUO
284402
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4G0G5
SCGB2B2
SCGBL Fusion Protein Ag15610
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SCGBL Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated