Learn More
SDC4 Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SDC4 |
---|---|
Concentration | 600 ÎĽg/mL |
Applications | Western Blot, Immunohistochemistry, Immunofluorescence, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899143
|
Proteintech
11820-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889143
|
Proteintech
11820-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SDC4, also named as syndecan 4, is a cell surface proteoglycan that bears heparan sulfate, which is involved in normal wound healing. It has been reported that SDC4 may be associated with keratinocyte proliferation and differentiation in the epidermis. SDC4 also plays a key role in the angiogenesis and inflammation. The predicted MW of SDC4 is 22 kDa and 11820-1-AP antibody recognizes a protein around 24-26 kDa and 48-52 kDa (homodimer) which is similar to the papers.(PMID: 20639201, 21949132; 18638267; 30165731 )
Specifications
SDC4 | |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
Unconjugated | |
Rabbit | |
6385 |
600 ÎĽg/mL | |
Polyclonal | |
Liquid | |
Human, Mouse | |
Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.