missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SDCCAG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15969-1-AP-20UL
This item is not returnable.
View return policy
Description
SDCCAG3, also named as NY-CO-3, is identified as serologically defined colon cancer antigen-3. It is an endosomal protein which is involved in the regulation of cytokinesis. SDCCAG3 is involved in modulation of TNF response. This gene has four isoforms with MW 40-50 kDa.Specifications
SDCCAG3 | |
Polyclonal | |
Unconjugated | |
SDCCAG3 | |
Antigen NY CO 3, NY CO 3, SDCCAG3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10807, 306322, 68112 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A2AIW0, Q96C92 | |
Sdccag3 | |
SDCCAG3 Fusion Protein Ag8745 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SDCCAG3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction