Learn More
SDHB Mouse anti-Human, Unconjugated, Clone: 1A2A5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67600-1-IG-20UL
Description
The SDHB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SDHB. This antibody recognizes human, mouse, rat antigen. The SDHB antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
SDHB | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
1A2A5 | |
Unconjugated | |
20 μL | |
6390 | |
IgG1 |