Learn More
SEC14L1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 25541-1-AP-150UL
 View more versions of this product
Product Code. 16894105
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
SEC14L1 may play a role in innate immunity by inhibiting the antiviral RIG-I signaling pathway. In this pathway, functions as a negative regulator of DDX58/RIG-I, the cytoplasmic sensor of viral nucleic acids. Prevents the interaction of DDX58 with MAVS/IPS1, an important step in signal propagation. May also regulate the SLC18A3 and SLC5A7 cholinergic transporters.
Specifications
Specifications
SEC14L1
Polyclonal
Unconjugated
SEC14L1
PRELID4A, SEC14 like 1 (S. cerevisiae), SEC14 like protein 1, SEC14L, SEC14L1
Rabbit
Antigen Affinity Chromatography
RUO
6397
-20°C
Liquid
Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q92503
SEC14L1
SEC14L1 Fusion Protein Ag22233
150 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SEC14L1 Rabbit anti-Human, Polyclonal, Proteintech > 150 μL; Unconjugated