Learn More
Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL Product Code.: 11327947
Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL
Product Code. 11327947
100 mL, 100mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Unit Size:
100mL
500mL
50mL
Product Code. 11327947
Brand: Thermo Scientific Alfa Aesar 014381.AE
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
MDL Number MFCD00134090
Specifications
Physical Form Liquid
Concentration Se in 5% HNO3
UN Number UN3264
Quantity 100 mL
Grade Specpure™
Chemical Name or Material Selenium plasma standard solution, Se 10000ÎĽg/mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number1534395 Certificate TypeCertificate of Analysis Date14/03/2025 Product Code014381.AP,014381.AE,014381.AD
Lot Number1517333 Certificate TypeCertificate of Analysis Date15/01/2025 Product Code014381.AP,014381.AE,014381.AD
Lot Number1510691 Certificate TypeCertificate of Analysis Date20/12/2024 Product Code014381.AP,014381.AE,014381.AD
Lot Number1490111 Certificate TypeCertificate of Analysis Date15/10/2024 Product Code014381.AP,014381.AE,014381.AD
Lot Number1477671 Certificate TypeCertificate of Analysis Date12/08/2024 Product Code014381.AP,014381.AE,014381.AD
RUO – Research Use Only
Product Content Correction
Product Title
Selenium, plasma standard solution, Specpure™ Se 10,000μg/mL >