missing translation for 'onlineSavingsMsg'
Learn More

SEMA4F Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1355.00 SEK - 3665.00 SEK

Specifications

Antigen SEMA4F
Concentration 0.27 mg/mL
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16840204
View Documents Promotion Details
Proteintech
12474-1-AP-150UL
150 μL
3665.00 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 06-12-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16850204
View Documents Promotion Details
Proteintech
12474-1-AP-20UL
20 μL
1355.00 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 06-12-2022
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SEMA4F has growth cone collapse activity against retinal ganglion-cell axons.
Specifications

Specifications

SEMA4F
Western Blot, Immunoprecipitation
Unconjugated
Rabbit
Mouse, Rat, Human
O95754, Q9Z123, Q9Z143
10505, 20355, 29745
SEMA4F Fusion Protein Ag3082
Primary
-20°C
SEMA4F
0.27 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
M SEMA, m Sema M, PRO2353, Sema M, Sema W, SEMA4F, SEMAM, Semaphorin 4F, Semaphorin M, Semaphorin W, SEMAW
SEMA4F
IgG
Antigen Affinity Chromatography
Antibody
SDS
Product Certifications