Learn More
Thermo Scientific Chemicals Semi dry blot transfer buffer (10X)
789.00 SEK - 1400.00 SEK
Specifications
Form Liquid
Storage Requirements Ambient temperatures
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15475839
View Documents
Thermo Scientific Chemicals
J63664.K2
1L
789.00 SEK
1 L
15485839
View Documents
Thermo Scientific Chemicals
J63664.K3
2L
1400.00 SEK
2 L
Description
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Liquid
Ambient temperatures
Documents
Product Certifications
RUO – Research Use Only
Product Content Correction
Product Title
Thermo Scientific Chemicals Semi dry blot transfer buffer (10X)