missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SENP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17659-1-AP-150UL
This item is not returnable.
View return policy
Description
The reversible posttranslational modification of proteins by the addition of small ubiquitin-like SUMO proteins is required for numerous biologic processes. SUMO-specific proteases, such as SENP3, are responsible for the initial processing of SUMO precursors to generate a C-terminal diglycine motif required for the conjugation reaction. They also have isopeptidase activity for the removal of SUMO from high molecular mass SUMO conjugates.Specifications
SENP3 | |
Polyclonal | |
Unconjugated | |
SENP3 | |
SENP3, Sentrin specific protease 3, SMT3IP1, SSP3, SUMO 1 specific protease 3, SUSP3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26168, 303245, 80886 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9EP97, Q9H4L4 | |
Senp3 | |
SENP3 Fusion Protein Ag11911 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SENP3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction