missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SEPT4 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890204
|
Proteintech
12476-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880204
|
Proteintech
12476-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Apoptosis is related to many diseases and development. Mitochondrial proteins, such as cytochrome c, Apaf-1, and AIF play important role in apoptosis. A novel mitochondrial septin-like protein was identified recently and designated ARTS for apoptosis related protein in TGF-beta signaling pathway. ARTS that is encoded by the human septin H5/PNUTL2/CDCrel2b gene is located to mitochondria and translocates to the nucleus when apoptosis occurs. ARTS is expressed in many tissues. It enhances cell death induced by TGF-beta and, to a lesser extent, by other apoptotic agents, such as TNF-a and Fas ligand.Specifications
SEPT4 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human | |
O43236 | |
5414 | |
SEPT4 Fusion Protein Ag3185 | |
Primary | |
-20°C | |
SEPT4 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
ARTS, BRADEION, Bradeion beta, Brain protein H5, CE5B3, CE5B3 beta, Cerebral protein 7, H5, hCDCREL 2, hucep 7, MART, Peanut like protein 2, PNUTL2, SEP4, SEPT4, septin 4 | |
SEPT4 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SEPT4 Rabbit anti-Human, Polyclonal, Proteintech