missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13818-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.Specifications
SEPT7 | |
Polyclonal | |
Unconjugated | |
SEPT7 | |
CDC10, CDC10 protein homolog, CDC3, Nbla02942, SEPT7, SEPT7A, septin 7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
235072, 64551, 989 | |
-20°C | |
Liquid |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O55131, Q16181, Q9WVC0 | |
SEPT7 | |
SEPT7 Fusion Protein Ag4907 | |
20 μL | |
Primary | |
Mouse, Rat, Human | |
Antibody | |
IgG |