missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | SEPT7 |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832214
|
Proteintech
13818-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16822214
|
Proteintech
13818-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 07-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a member of the Jumonji, AT-rich interactive domain 1 (JARID1) histone demethylase protein family. The encoded protein plays a role in gene regulation through the histone code by specifically demethylating lysine 4 of histone H3. The encoded protein interacts with many other proteins, including retinoblastoma protein, and is implicated in the transcriptional regulation of Hox genes and cytokines. This gene may play a role in tumor progression.Specifications
SEPT7 | |
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
O55131, Q16181, Q9WVC0 | |
235072, 64551, 989 | |
SEPT7 Fusion Protein Ag4907 | |
Primary | |
-20°C | |
SEPT7 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CDC10, CDC10 protein homolog, CDC3, Nbla02942, SEPT7, SEPT7A, septin 7 | |
SEPT7 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |