Learn More
Robu™ Series 16 Biplane Filter Discs
Robu™ Series 16 Biplane Filter Discs are precisely ground down to tight tolerances and plane surfaces, which may be required in specific installation circumstances.
Brand: Robu™ 16303
Additional Details : Weight : 0.04940kg
Description
- ISO 4793-80
- Surfaces plane
- Ground cylindrical
- Faces are precisely ground down to tight tolerances and plane surfaces, which might be required in specific installation circumstances
Specifications
Biplane | |
Cylindrical | |
Filter Disc |
16 to 40 μm | |
3 ± 0.2 mm | |
30 mm |