Learn More
Robu™ Series 16 Biplane Filter Discs
Robu™ Series 16 Biplane Filter Discs are precisely ground down to tight tolerances and plane surfaces, which may be required in specific installation circumstances.
875.00 SEK - 900.00 SEK
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Membrane | Price | Quantity & Availability | |||||
16852836
|
Robu™
16303 |
30 mm |
875.00 SEK
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
16882826
|
Robu™
16254 |
25 mm |
900.00 SEK
Pack of 10 |
Estimated Shipment: 29-05-2024
Log in to see stock. |
|||||
Description
- ISO 4793-80
- Surfaces plane
- Ground cylindrical
- Faces are precisely ground down to tight tolerances and plane surfaces, which might be required in specific installation circumstances
Specifications
Biplane | |
Cylindrical | |
Filter Disc |