Learn More
Robu™ Series 16 Biplane Filter Discs
Robu™ Series 16 Biplane Filter Discs are precisely ground down to tight tolerances and plane surfaces, which may be required in specific installation circumstances.
Brand:  Robu™ 16254
Additional Details : Weight : 0.03230kg
 View more versions of this product
Product Code. 16882826
  • 900.00 SEK / Pack of 10
Estimated Shipment: 29-05-2024
to see stock.
Add to basket
Description
Description
  • ISO 4793-80
  • Surfaces plane
  • Ground cylindrical
  • Faces are precisely ground down to tight tolerances and plane surfaces, which might be required in specific installation circumstances
Specifications
Specifications
Biplane
Cylindrical
Filter Disc
10 to 16 μm
2.5 ± 0.2 mm
25 mm