missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | SERPINA10 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16899193
|
Proteintech
11851-1-AP-20UL |
20 μL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16889193
|
Proteintech
11851-1-AP-150UL |
150 μL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene belongs to the serpin family. It is predomitly expressed in the liver and secreted in plasma. It inhibits the activity of coagulation factors Xa and XIa in the presence of protein Z, calcium and phospholipid. Mutations in this gene are associated with venous thrombosis. Alternatively spliced transcript variants have been found for this gene.Specifications
SERPINA10 | |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q62975, Q8R121, Q9UK55 | |
171154, 217847, 51156 | |
SERPINA10 Fusion Protein Ag2420 | |
Primary | |
-20°C | |
SERPINA10 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
PZI, Serpin A10, SERPINA10, ZPI | |
Serpina10 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SERPINA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech