missing translation for 'onlineSavingsMsg'
Learn More

Serum amyloid P component Mouse anti-Human, Clone: 1A1D10, Proteintech

Mouse Monoclonal Antibody

1645.54 SEK - 4652.90 SEK

Specifications

Antigen Serum amyloid P component
Clone 1A1D10
Concentration 0.42 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897635
View Documents
Proteintech
66084-1-IG-20UL
20 ÎĽL
1645.54 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16887635
View Documents
Proteintech
66084-1-IG-150UL
150 ÎĽL
4652.90 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The protein encoded by this gene is a glycoprotein, belonging to the pentraxin family of proteins, which has a characteristic pentameric organization. These family members have considerable sequence homology which is thought to be the result of gene duplication. The binding of the encoded protein to proteins in the pathological amyloid cross-beta fold suggests its possible role as a chaperone. This protein is also thought to control the degradation of chromatin. It has been demonstrated that this protein binds to apoptotic cells at an early stage, which raises the possibility that it is involved in dealing with apoptotic cells in vivo.
Specifications

Specifications

Serum amyloid P component
0.42 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
9.5S alpha 1 glycoprotein, amyloid P component, serum, APCS, PTX2, SAP, Serum amyloid P component
APCS
IgG2b
Protein A
Antibody
1A1D10
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
P02743
325
Serum amyloid P component Fusion Protein Ag14957
Primary
-20°C
APCS
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Serum amyloid P component Mouse anti-Human, Clone: 1A1D10, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.