Learn More
SF3A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12070-1-AP-20UL
 View more versions of this product
Product Code. 16879563
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
This gene encodes subunit 3 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 3 interacts with subunit 1 through its amino-terminus while the zinc finger domain of subunit 3 plays a role in its binding to the 15S U2 snRNP. This gene has a pseudogene on chromosome 20. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
SF3A3
Polyclonal
Unconjugated
SF3A3
PRP9, PRPF9, SAP 61, SAP61, SF3A3, SF3a60, Splicing factor 3A subunit 3
Rabbit
Antigen Affinity Chromatography
RUO
10946, 313583, 75062
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Immunocytochemistry
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q12874, Q9D554
Sf3a3
SF3A3 Fusion Protein Ag2708
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SF3A3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated