Learn More
Trajan™ Fixed Luer Tip Syringes Product Code.: 10452571
Trajan™ Fixed Luer Tip Syringes
Product Code. 10452571
Each
Click to view available options
Unit Size:
Each
Product Code. 10452571
Brand: Trajan™ 005229
Additional replacement needles for Luer Tip Gas Tight Syringe.
  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Manual syringe
Specifications
Material Borosilicate Glass
For Use With (Application) For maximum inertness
Product Type Manual Syringe
Volume (Metric) 100 ÎĽL
Connection Type Luer Tip, Fixed
Product Content Correction
Product Title
Trajan™ Fixed Luer Tip Syringes >