Learn More
Trajan™ Replacement Needles for 5μL eVol™ NMR Syringes
For gas chromatography systems
Brand:  Trajan™ 036910
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 12907606
  • 600.00 SEK / Pack of 5
Estimated Shipment: 20-06-2024
to see stock.
Description
Description
  • Needle length is the exposed length of needle
  • Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Specifications
Stainless Steel
0.004 in.
0.11 mm
2 in.
50 mm
eVol NMR Syringe (5μL)
25 G
0.019 in.
0.5 mm
Replacement Needle
Bevel