Learn More
Trajan™ Replacement Needles for 5μL eVol™ NMR Syringes
For gas chromatography systems
555.00 SEK - 600.00 SEK
Specifications
Material | Stainless Steel |
---|---|
Needle Gauge | 25 G |
Diameter (English) Outer | 0.019 in. |
Diameter (Metric) Outer | 0.5 mm |
Product Type | Replacement Needle |
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) Needle | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Length (Metric) Needle | Price | Quantity & Availability | ||||
12907606
|
Trajan™
036910 |
0.11 mm | 50 mm |
600.00 SEK
Pack of 5 |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12967606
|
Trajan™
036920 |
0.2 mm | 115 mm |
555.00 SEK
Pack of 2 |
Estimated Shipment: 10-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Needle length is the exposed length of needle
- Choose a different replacement needle by selecting syringe volume, needle length, OD and tip style
Specifications
Stainless Steel | |
0.019 in. | |
Replacement Needle | |
eVol NMR Syringe (5μL) |
25 G | |
0.5 mm | |
Bevel |