Learn More
SGE™ SilFlow™ GC Replacement Parts
Tightened using finger force to achieve a reliable seal, even for sensitive MS systems
Brand: SGE™ 123704
Additional Details : Weight : 0.01000kg
Description
- Conventional graphite or Vespel™ ferrules cannot be used with the SilFlow MCD as the dimensional stability is not adequate and there is a risk of contaminating the channels
- SilTite™ FingerTite metal ferrules result in a reliable zero dead volume connection
Specifications
SGE Silflow Microchannel Device | |
Nut | |
— | |
Nut |