Learn More
Trajan™ Syringe with Valve
Ideal for sample storage and transportation or to pressurize sample prior to injection.
Brand: Trajan™ 004279
Additional Details : Weight : 0.01000kg
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Pre-fitted with a valve for sample pressurization prior to injection
Specifications
PTFE (Plunger) | |
Pre-fitted with Syringe Valves | |
50 μL |