missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGF29 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24061-1-AP-20UL
This item is not returnable.
View return policy
Description
CCDC101 is a subunit of 2 histone acetyltransferase complexes: the ADA2A -containing -TAF9 /PCAF acetylase (STAGA) complex. Both of these complexes contain either GCN5 or PCAF, which are paralogous acetyltransferases.Specifications
SGF29 | |
Polyclonal | |
Unconjugated | |
SGF29 | |
CCDC101, SGF29 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
112869 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96ES7 | |
SGF29 | |
SGF29 Fusion Protein Ag21261 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SGF29 Rabbit anti-Human, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction